5-Bromo-1,3-dichloro-2-(difluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

37026-500MG 37026-1G 37026-5G
77326-268EA 348.14 USD
77326-268 77349-976 77374-194
5-Bromo-1,3-dichloro-2-(difluoromethoxy)benzene ≥97%
5-Bromo-1,3-dichloro-2-(difluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₃BrCl₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD22580832
CAS Number: 929621-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR