(3,4-Dichloro-2-fluorophenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

37016-500MG 37016-1G 37016-5G 37016-25G
77326-264EA 244.11 USD
77326-264 77349-970 77362-580 77385-456
(3,4-Dichloro-2-fluorophenyl)methanol ≥95%
(3,4-Dichloro-2-fluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₅Cl₂FO
Storage Temperature: Ambient
MDL Number: MFCD28786032
CAS Number: 1615212-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR