1-Bromo-2,3-difluoro-4-(methoxymethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

37015-500MG 37015-1G 37015-5G
77336-788EA 394.49 USD
77336-788 77349-968 77374-188
1-Bromo-2,3-difluoro-4-(methoxymethyl)benzene ≥95%
1-Bromo-2,3-difluoro-4-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₇BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD30646485
CAS Number: 162744-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR