5-Chloro-2-(methoxymethyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

37013-500MG 37013-1G 37013-25G 37013-5G
77326-262EA 306.94 USD
77326-262 77349-966 77385-454 77374-186
5-Chloro-2-(methoxymethyl)pyridine ≥95%
5-Chloro-2-(methoxymethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₈ClNO
MW: 157.598 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28130259
CAS Number: 1446105-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR