3-Fluoro-4-(methoxymethyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

37012-500MG 37012-1G 37012-5G 37012-25G
77326-260EA 95.79 USD
77326-260 77342-544 77362-578 77374-184
3-Fluoro-4-(methoxymethyl)pyridine ≥95%
3-Fluoro-4-(methoxymethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₈FNO
MW: 141.143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632130
CAS Number: 2379321-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR