3,5-Dichloro-4-(methoxymethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

37006-500MG 37006-1G 37006-5G
77335-794EA 366.68 USD
77335-794 77349-964 77374-182
3,5-Dichloro-4-(methoxymethyl)pyridine ≥97%
3,5-Dichloro-4-(methoxymethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₇Cl₂NO
MW: 192.043 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632128
CAS Number: 2379321-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR