5-Bromo-2-ethoxy-4-(methoxymethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

37001-500MG 37001-1G 37001-5G
77335-390EA 353.29 USD
77335-390 77349-960 77374-178
5-Bromo-2-ethoxy-4-(methoxymethyl)pyridine ≥97%
5-Bromo-2-ethoxy-4-(methoxymethyl)pyridine

Precaución: for Research Use Only

Formula: C₉H₁₂BrNO₂
MW: 246.101 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632127
CAS Number: 2379322-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR