5-Bromo-4-(methoxymethyl)benzo[d][1,3]dioxole ≥95%

Supplier: AOBChem USA

Warning

36993-500MG 36993-1G 36993-5G 36993-25G
77326-256EA 144.2 USD
77326-256 77345-050 77362-574 77385-450
5-Bromo-4-(methoxymethyl)benzo[d][1,3]dioxole ≥95%
5-Bromo-4-(methoxymethyl)benzo[d][1,3]dioxole

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD32632124
CAS Number: 669051-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR