2-(Difluoromethoxy)-1-fluoro-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

36986-500MG 36986-1G 36986-5G
77338-566EA 498.52 USD
77338-566 77349-958 77374-176
2-(Difluoromethoxy)-1-fluoro-3-(trifluoromethyl)benzene ≥97%
2-(Difluoromethoxy)-1-fluoro-3-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₄F₆O
MW: 230.107 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28789462
CAS Number: 1803848-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR