1-(Benzyloxy)-4-bromo-2-chloro-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

36983-500MG 36983-1G 36983-5G 36983-25G
77326-254EA 146.26 USD
77326-254 77345-152 77362-572 77385-448
1-(Benzyloxy)-4-bromo-2-chloro-3-methylbenzene ≥95%
1-(Benzyloxy)-4-bromo-2-chloro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrClO
MW: 311.601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632121
CAS Number: 2379321-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR