1-(3-Bromo-4-methylphenyl)-2,5-dimethyl-1H-pyrrole ≥97%
Supplier: AOBChem USA
|
Warning
|
36970-500MG
36970-1G
36970-5G
36970-25G
77326-250EA
168.92
USD
77326-250
77346-316
77362-570
77385-446
1-(3-Bromo-4-methylphenyl)-2,5-dimethyl-1H-pyrrole ≥97%
1-(3-Bromo-4-methylphenyl)-2,5-dimethyl-1H-pyrrole
Precaución: for Research Use Only
Formula:
C₁₃H₁₄BrN MW: 264.161 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08671318 CAS Number: 736973-12-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...