2-(Benzyloxy)-1-fluoro-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

36969-1G 36969-5G 36969-25G
77341-302EA 82.4 USD
77341-302 77362-568 77374-172
2-(Benzyloxy)-1-fluoro-3-(trifluoromethyl)benzene ≥97%
2-(Benzyloxy)-1-fluoro-3-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₄O
MW: 270.222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632118
CAS Number: 2379322-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR