1-Chloro-2-ethoxy-4-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

36924-1G 36924-5G 36924-25G
77341-102EA 71.07 USD
77341-102 77362-564 77374-166
1-Chloro-2-ethoxy-4-methoxybenzene ≥95%
1-Chloro-2-ethoxy-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₁ClO₂
MW: 186.635 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632112
CAS Number: 2379322-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR