2-Bromo-6-fluoro-3-hydroxybenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
36922-500MG
36922-1G
36922-5G
77337-538EA
418.18
USD
77337-538
77349-950
77374-164
2-Bromo-6-fluoro-3-hydroxybenzoic acid ≥97%
2-Bromo-6-fluoro-3-hydroxybenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₄BrFO₃ MW: 235.007 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28738973 CAS Number: 91659-32-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...