(2-(Benzyloxy)naphthalen-1-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

36909-500MG 36909-1G 36909-5G 36909-25G
77326-236EA 131.84 USD
77326-236 77344-370 77362-560 77385-442
(2-(Benzyloxy)naphthalen-1-yl)methanol ≥95%
(2-(Benzyloxy)naphthalen-1-yl)methanol

Precaución: for Research Use Only

Formula: C₁₈H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD04492344
CAS Number: 217176-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR