6-(Benzyloxy)-5-chloro-2-naphthoic acid ≥97%

Supplier: AOBChem USA

Warning

36906-500MG 36906-1G 36906-5G
77339-204EA 564.44 USD
77339-204 77349-942 77374-154
6-(Benzyloxy)-5-chloro-2-naphthoic acid ≥97%
6-(Benzyloxy)-5-chloro-2-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₈H₁₃ClO₃
MW: 312.747 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756032
CAS Number: 2764729-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR