(3-(Benzyloxy)-6-bromo-2-fluorophenyl)(methyl)sulfane ≥97%
Supplier: AOBChem USA
|
Warning
|
36899-500MG
36899-1G
36899-5G
36899-25G
77326-232EA
131.84
USD
77326-232
77344-368
77362-558
77385-440
(3-(Benzyloxy)-6-bromo-2-fluorophenyl)(methyl)sulfane ≥97%
(3-(Benzyloxy)-6-bromo-2-fluorophenyl)(methyl)sulfane
Precaución: for Research Use Only
Formula:
C₁₄H₁₂BrFOS Storage Temperature: Ambient |
MDL Number:
MFCD32632102 CAS Number: 2379322-09-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...