1-(Benzyloxy)-4-bromo-3-ethoxy-2-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

36892-500MG 36892-1G 36892-5G 36892-25G
77326-230EA 218.36 USD
77326-230 77349-936 77362-556 77385-438
1-(Benzyloxy)-4-bromo-3-ethoxy-2-fluorobenzene ≥97%
1-(Benzyloxy)-4-bromo-3-ethoxy-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₄BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD32632101
CAS Number: 2379322-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR