(((4-Bromo-2-fluoro-1,3-phenylene)bis(oxy))bis(methylene))dibenzene ≥97%

Supplier: AOBChem USA

Warning

36891-500MG 36891-1G 36891-5G 36891-25G
77326-228EA 218.36 USD
77326-228 77349-934 77362-554 77385-436
(((4-Bromo-2-fluoro-1,3-phenylene)bis(oxy))bis(methylene))dibenzene ≥97%
(((4-Bromo-2-fluoro-1,3-phenylene)bis(oxy))bis(methylene))dibenzene

Precaución: for Research Use Only

Formula: C₂₀H₁₆BrFO₂
MW: 387.242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632100
CAS Number: 2379322-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR