Methyl 4-(difluoromethoxy)-1-naphthoate ≥97%

Supplier: AOBChem USA

Warning

36874-500MG 36874-1G 36874-5G
77338-010EA 458.35 USD
77338-010 77349-930 77374-146
Methyl 4-(difluoromethoxy)-1-naphthoate ≥97%
Methyl 4-(difluoromethoxy)-1-naphthoate

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₂O₃
MW: 252.214 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18414961
CAS Number: 1261559-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR