4-(Difluoromethoxy)-1-naphthoic acid ≥97%

Supplier: AOBChem USA

Warning

36873-500MG 36873-1G 36873-5G
77336-786EA 394.49 USD
77336-786 77349-928 77374-144
4-(Difluoromethoxy)-1-naphthoic acid ≥97%
4-(Difluoromethoxy)-1-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₂H₈F₂O₃
MW: 238.187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18414093
CAS Number: 1261884-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR