(4-(Difluoromethoxy)naphthalen-1-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

36864-500MG 36864-1G 36864-5G
77340-122EA 767.35 USD
77340-122 77349-926 77374-140
(4-(Difluoromethoxy)naphthalen-1-yl)methanol ≥95%
(4-(Difluoromethoxy)naphthalen-1-yl)methanol

Precaución: for Research Use Only

Formula: C₁₂H₁₀F₂O₂
MW: 224.203 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18414105
CAS Number: 668275-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR