4-(Difluoromethoxy)-1-naphthaldehyde ≥95%

Supplier: AOBChem USA

Warning

36863-500MG 36863-1G 36863-5G
77339-586EA 602.55 USD
77339-586 77349-924 77374-138
4-(Difluoromethoxy)-1-naphthaldehyde ≥95%
4-(Difluoromethoxy)-1-naphthaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18414081
CAS Number: 842173-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR