2-(Benzo[d]thiazol-2-yl)-5-bromophenol ≥95%

Supplier: AOBChem USA

Warning

36810-500MG 36810-1G 36810-5G 36810-25G
77326-218EA 161.71 USD
77326-218 77345-850 77362-544 77385-430
2-(Benzo[d]thiazol-2-yl)-5-bromophenol ≥95%
2-(Benzo[d]thiazol-2-yl)-5-bromophenol

Precaución: for Research Use Only

Formula: C₁₃H₈BrNOS
MW: 306.178 g/mol
Storage Temperature: Ambient
CAS Number: 90481-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR