1,2,5-Tribromo-4-chloro-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

36796-500MG 36796-1G 36796-5G 36796-25G
77326-216EA 195.7 USD
77326-216 77349-918 77362-540 77385-428
1,2,5-Tribromo-4-chloro-3-methylbenzene ≥95%
1,2,5-Tribromo-4-chloro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₄Br₃Cl
MW: 363.272 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632086
CAS Number: 2379321-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR