4-Chloro-1-fluoro-2-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

36789-500MG 36789-1G 36789-5G 36789-25G
77326-214EA 121.54 USD
77326-214 77343-908 77362-538 77374-130
4-Chloro-1-fluoro-2-(methoxymethoxy)benzene ≥95%
4-Chloro-1-fluoro-2-(methoxymethoxy)benzene

Precaución: for Research Use Only

Formula: C₈H₈ClFO₂
MW: 190.599 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632085
CAS Number: 1865062-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR