3'-Methyl-[1,1'-biphenyl]-4-carbonitrile ≥97%

Supplier: AOBChem USA

Warning

36770-1G 36770-5G 36770-25G
77341-100EA 71.07 USD
77341-100 77362-536 77374-124
3'-Methyl-[1,1'-biphenyl]-4-carbonitrile ≥97%
3'-Methyl-[1,1'-biphenyl]-4-carbonitrile

Precaución: for Research Use Only

Formula: C₁₄H₁₁N
MW: 193.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05980908
CAS Number: 132312-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR