1-(Benzyloxy)-4-bromo-5-fluoro-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

36763-1G 36763-5G 36763-25G
77340-650EA 60.77 USD
77340-650 77362-532 77374-122
1-(Benzyloxy)-4-bromo-5-fluoro-2-methoxybenzene ≥97%
1-(Benzyloxy)-4-bromo-5-fluoro-2-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFO₂
MW: 311.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28403568
CAS Number: 1049093-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR