2-Isopropoxy-5-(methoxymethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

36755-500MG 36755-1G 36755-5G 36755-25G
77326-206EA 193.64 USD
77326-206 77349-910 77362-530 77385-422
2-Isopropoxy-5-(methoxymethyl)pyridine ≥97%
2-Isopropoxy-5-(methoxymethyl)pyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₂
MW: 181.232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632080
CAS Number: 2379321-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR