6-(Difluoromethoxy)-2-naphthoic acid ≥97%

Supplier: AOBChem USA

Warning

36742-500MG 36742-1G 36742-5G
77339-180EA 560.32 USD
77339-180 77349-908 77374-118
6-(Difluoromethoxy)-2-naphthoic acid ≥97%
6-(Difluoromethoxy)-2-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₂H₈F₂O₃
MW: 238.187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18414100
CAS Number: 1261884-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR