3-Bromo-2-(difluoromethoxy)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

36739-500MG 36739-1G 36739-5G
77338-980EA 539.72 USD
77338-980 77349-906 77374-116
3-Bromo-2-(difluoromethoxy)benzoic acid ≥95%
3-Bromo-2-(difluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD14656706
CAS Number: 1249235-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR