1-Bromo-4-fluoro-5-(methoxymethyl)-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

36733-500MG 36733-1G 36733-5G 36733-25G
77326-204EA 109.18 USD
77326-204 77343-238 77362-528 77374-114
1-Bromo-4-fluoro-5-(methoxymethyl)-2-methylbenzene ≥97%
1-Bromo-4-fluoro-5-(methoxymethyl)-2-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO
MW: 233.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632076
CAS Number: 2379322-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR