1-Chloro-2-(methoxymethyl)-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

36732-500MG 36732-1G 36732-5G 36732-25G
77326-202EA 136.99 USD
77326-202 77344-524 77362-526 77385-420
1-Chloro-2-(methoxymethyl)-3-(trifluoromethyl)benzene ≥97%
1-Chloro-2-(methoxymethyl)-3-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₈ClF₃O
MW: 224.607 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632075
CAS Number: 2379321-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR