(3-Bromo-2-isopropoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

36727-500MG 36727-1G 36727-5G
77326-200EA 317.24 USD
77326-200 77349-902 77374-110
(3-Bromo-2-isopropoxyphenyl)methanol ≥95%
(3-Bromo-2-isopropoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO₂
MW: 245.113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14656747
CAS Number: 1250782-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR