Methyl 2-(benzyloxy)-3-fluorobenzoate ≥95%

Supplier: AOBChem USA

Warning

36714-500MG 36714-1G 36714-5G 36714-25G
77326-196EA 218.36 USD
77326-196 77349-900 77362-522 77385-418
Methyl 2-(benzyloxy)-3-fluorobenzoate ≥95%
Methyl 2-(benzyloxy)-3-fluorobenzoate

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₃
MW: 260.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632067
CAS Number: 2379322-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR