2-Bromo-N,N-dimethyl-5-(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

36712-1G 36712-5G 36712-25G
77341-752EA 104.03 USD
77341-752 77362-518 77374-106
2-Bromo-N,N-dimethyl-5-(trifluoromethyl)benzamide ≥97%
2-Bromo-N,N-dimethyl-5-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₀H₉BrF₃NO
MW: 296.084 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25132880
CAS Number: 1369904-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR