(2-Bromo-5-(trifluoromethyl)phenyl)(pyrrolidin-1-yl)methanone ≥97%
Supplier: AOBChem USA
|
Warning
|
36711-1G
36711-5G
36711-25G
77342-074EA
112.27
USD
77342-074
77362-516
77374-104
(2-Bromo-5-(trifluoromethyl)phenyl)(pyrrolidin-1-yl)methanone ≥97%
(2-Bromo-5-(trifluoromethyl)phenyl)(pyrrolidin-1-yl)methanone
Precaución: for Research Use Only
Formula:
C₁₂H₁₁BrF₃NO MW: 322.121 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD25132867 CAS Number: 1543076-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...