(2-Bromo-5-(trifluoromethyl)phenyl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

36710-1G 36710-5G 36710-25G
77342-072EA 112.27 USD
77342-072 77362-514 77374-102
(2-Bromo-5-(trifluoromethyl)phenyl)(thiomorpholino)methanone ≥97%
(2-Bromo-5-(trifluoromethyl)phenyl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₁BrF₃NOS
MW: 354.186 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25624409
CAS Number: 1510883-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR