Ethyl 2-chloro-6-fluoro-3-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

36700-500MG 36700-1G 36700-5G 36700-25G
77326-192EA 119.48 USD
77326-192 77343-638 77362-512 77374-100
Ethyl 2-chloro-6-fluoro-3-methoxybenzoate ≥97%
Ethyl 2-chloro-6-fluoro-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD10566181
CAS Number: 1397199-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR