tert-Butyl 2-chloro-6-fluoro-3-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

36699-1G 36699-5G 36699-25G
77341-098EA 71.07 USD
77341-098 77362-510 77374-098
tert-Butyl 2-chloro-6-fluoro-3-methoxybenzoate ≥97%
tert-Butyl 2-chloro-6-fluoro-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₄ClFO₃
MW: 260.689 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632062
CAS Number: 2379322-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR