2-(Difluoromethoxy)-4-fluoro-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

36690-500MG 36690-1G 36690-5G 36690-25G
77326-186EA 263.68 USD
77326-186 77349-896 77362-504 77385-412
2-(Difluoromethoxy)-4-fluoro-1-methylbenzene ≥97%
2-(Difluoromethoxy)-4-fluoro-1-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₇F₃O
MW: 176.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698456
CAS Number: 1214333-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR