Methyl-6-methoxy-2-naphthoate ≥97%

Supplier: AOBChem USA

Warning

36672-500MG 36672-1G 36672-5G 36672-25G
77326-184EA 106.09 USD
77326-184 77343-116 77362-502 77374-094
Methyl-6-methoxy-2-naphthoate ≥97%
Methyl-6-methoxy-2-naphthoate

Precaución: for Research Use Only

Formula: C₁₃H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD01108716
CAS Number: 5043-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR