4-Ethoxy-2,5-dimethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

36669-500MG 36669-1G 36669-5G
77339-646EA 606.67 USD
77339-646 77349-892 77374-088
4-Ethoxy-2,5-dimethoxybenzaldehyde ≥97%
4-Ethoxy-2,5-dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₄
MW: 210.226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19302344
CAS Number: 873984-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR