5-(Benzyloxy)-2-fluoronicotinic acid ≥97%

Supplier: AOBChem USA

Warning

36665-500MG 36665-1G 36665-5G
77338-330EA 477.92 USD
77338-330 77349-890 77374-086
5-(Benzyloxy)-2-fluoronicotinic acid ≥97%
5-(Benzyloxy)-2-fluoronicotinic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₀FNO₃
MW: 247.222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18255066
CAS Number: 1256790-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR