4-Bromo-2-fluoro-3-isopropoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

36662-500MG 36662-1G 36662-5G 36662-25G
77326-182EA 161.71 USD
77326-182 77345-848 77362-500 77385-410
4-Bromo-2-fluoro-3-isopropoxybenzaldehyde ≥97%
4-Bromo-2-fluoro-3-isopropoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrFO₂
MW: 261.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632055
CAS Number: 2379321-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR