2-Fluoro-3-methoxy-5-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

36656-500MG 36656-1G 36656-5G 36656-25G
77326-180EA 226.6 USD
77326-180 77349-888 77362-498 77385-408
2-Fluoro-3-methoxy-5-(methylthio)benzoic acid ≥97%
2-Fluoro-3-methoxy-5-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃S
MW: 216.229 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22490195
CAS Number: 1824583-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR