2-Bromo-3-fluoro-4-iodo-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

36653-500MG 36653-1G 36653-5G 36653-25G
77326-178EA 166.86 USD
77326-178 77346-194 77362-496 77385-406
2-Bromo-3-fluoro-4-iodo-1-methoxybenzene ≥97%
2-Bromo-3-fluoro-4-iodo-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅BrFIO
MW: 330.921 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756025
CAS Number: 2387195-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR