4-(Benzyloxy)-2-(methoxymethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

36647-500MG 36647-1G 36647-5G 36647-25G
77326-174EA 243.08 USD
77326-174 77349-884 77362-494 77385-404
4-(Benzyloxy)-2-(methoxymethyl)benzaldehyde ≥97%
4-(Benzyloxy)-2-(methoxymethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₃
MW: 256.296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632053
CAS Number: 2379322-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR