4-(Benzyloxy)-2-(methoxymethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

36646-500MG 36646-1G 36646-5G 36646-25G
77326-172EA 243.08 USD
77326-172 77349-882 77362-492 77385-402
4-(Benzyloxy)-2-(methoxymethyl)benzoic acid ≥97%
4-(Benzyloxy)-2-(methoxymethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₄
MW: 272.296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632052
CAS Number: 2379321-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR