2-Bromo-6-chloro-3-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

36611-500MG 36611-1G 36611-5G 36611-25G
77326-166EA 263.68 USD
77326-166 77349-872 77362-490 77385-398
2-Bromo-6-chloro-3-(methylthio)benzaldehyde ≥97%
2-Bromo-6-chloro-3-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrClOS
MW: 265.555 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403717
CAS Number: 2624417-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR